Assay Details
Gene Symbol: BAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204878.1 | NP_001191807.1 | 2 - 3 | 807 | 130 | |
NM_004874.3 | NP_004865.1 | 3 - 4 | 915 | 130 | ||
GenBank mRNA | AF095194.2 | - | 3 - 4 | 675 | 130 | |
AF111116.1 | - | 3 - 4 | 633 | 130 | ||
AK304072.1 | - | 2 - 3 | 584 | 130 | ||
AK314020.1 | - | 3 - 4 | 688 | 130 | ||
BC000800.1 | - | 2 - 3 | 807 | 130 | ||
BC038505.1 | - | 3 - 4 | 709 | 5259771 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
