Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027121.hg.1 |
ENST00000296980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027121.hg.1 |
ENST00000349184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027121.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027121.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027121.hg.1 |
TC06002151.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
ENST00000296980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
ENST00000349184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027122.hg.1 |
TC06002151.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013299.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
TC0600013299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
ENST00000339602.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192039.hg.1 |
NM_181310.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
NM_181309.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
TC0600013299.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
ENST00000296980.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
ENST00000349184.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
NM_052962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600192038.hg.1 |
NM_181309.1 |
Refseq |