Assay Details
Gene Symbol: KPNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012316.4 | NP_036448.1 | 7 - 8 | 738 | 86 | |
XM_005270711.3 | XP_005270768.1 | 7 - 8 | 1099 | 86 | ||
GenBank mRNA | AB621806.1 | - | 7 - 8 | 727 | 86 | |
AF060543.1 | - | 7 - 8 | 705 | 86 | ||
AK128609.1 | - | 7 - 8 | 701 | 86 | ||
AK295615.1 | - | 7 - 8 | 853 | 86 | ||
AK301714.1 | - | 7 - 8 | 692 | 86 | ||
AK315490.1 | - | 7 - 8 | 725 | 86 | ||
BC020520.1 | - | 7 - 8 | 713 | 3913864 | 86 | |
BT009843.1 | - | 6 - 7 | 650 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000419.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | ENST00000373617.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | ENST00000373625.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | ENST00000471599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | ENST00000537234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | ENST00000545542.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01006401.hg.1 | TC01000419.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | ENST00000515055.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007662.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | NM_012316.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | TC0100007662.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000373625.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000446515.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000469790.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | ENST00000471599.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100153798.hg.1 | NM_012316.4 | Refseq |
