Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003542.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003542.hg.1 |
ENST00000546171.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003542.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003542.hg.1 |
TC11000262.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149875.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100149875.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149875.hg.1 |
TC1100007064.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100149875.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100149875.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000263160.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
ENST00000534115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007064.hg.1 |
NM_020346.2 |
Refseq |