Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023922.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023922.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023922.hg.1 |
TC11001990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023924.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023924.hg.1 |
TC11001990.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100187506.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187506.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187506.hg.1 |
TC1100011344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100187506.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187506.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011344.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
NR_033791.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
TC1100011344.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
ENST00000495263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
ENST00000514950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187505.hg.1 |
NR_033791.1 |
Refseq |