Assay Details
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005727.3 | NP_005718.2 | 5 - 6 | 818 | 61 | |
XM_011540460.2 | XP_011538762.1 | 5 - 6 | 792 | 61 | ||
GenBank mRNA | AF054838.1 | - | 3 - 4 | 344 | 61 | |
AF065388.1 | - | 4 - 5 | 465 | 61 | ||
AF133425.1 | - | 4 - 5 | 467 | 61 | ||
AK313774.1 | - | 5 - 6 | 808 | 61 | ||
BC007290.1 | - | 4 - 5 | 471 | 3349295 | 61 | |
BC013404.2 | - | 4 - 5 | 469 | 2959379 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000580.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009864.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009864.hg.1 | ENST00000498443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009864.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009864.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158238.hg.1 | NM_005727.3 | Refseq |
