Assay Details
Gene Symbol: ATAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024857.4 | NP_079133.3 | 10 - 11 | 3515 | 131 | |
XM_011525269.2 | XP_011523571.1 | 10 - 11 | 3539 | 131 | ||
XM_011525270.2 | XP_011523572.1 | 10 - 11 | 3539 | 131 | ||
XM_011525274.2 | XP_011523576.1 | 5 - 6 | 767 | 131 | ||
XM_017025111.1 | XP_016880600.1 | 10 - 11 | 3514 | 131 | ||
GenBank mRNA | AJ314648.2 | - | 10 - 11 | 3483 | 131 | |
AK022797.1 | - | 9 - 10 | 2010 | 131 | ||
AL832103.1 | - | 10 - 11 | 3516 | 131 | ||
AY557611.1 | - | 10 - 11 | 3483 | 131 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000355.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17004451.hg.1 | ENST00000321990.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17004451.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17004451.hg.1 | TC17000355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17004450.hg.1 | ENST00000321990.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17004450.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17004450.hg.1 | TC17000355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17004452.hg.1 | ENST00000321990.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17004452.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17004452.hg.1 | TC17000355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000390888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | TC1700007469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153487.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000390888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | TC1700007469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153489.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | TC1700007469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153491.hg.1 | NM_024857.4 | Refseq |
