Assay Details
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003655.2 | NP_003646.2 | 1 - 2 | 233 | 67 | |
GenBank mRNA | AF013956.1 | - | 1 - 2 | 236 | 67 | |
AY390430.1 | - | 1 - 2 | 70 | 67 | ||
BC014967.1 | - | 1 - 2 | 89 | 4856728 | 67 | |
BG029608.1 | - | 1 - 2 | 66 | 67 | ||
BQ225831.1 | - | 233 | 67 | |||
EU439707.1 | - | 1 - 2 | 265 | 67 | ||
KU178422.1 | - | 1 - 2 | 70 | 67 | ||
KU178423.1 | - | 1 - 2 | 70 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001930.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
protein sumoylation
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
transcription cis-regulatory region binding
chromatin binding
transcription corepressor activity
single-stranded RNA binding
protein binding
ligase activity
SUMO transferase activity
enzyme binding
SUMO binding
methylated histone binding
phosphoprotein binding
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
protein sumoylation
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
transcription cis-regulatory region binding
chromatin binding
transcription corepressor activity
single-stranded RNA binding
protein binding
ligase activity
SUMO transferase activity
enzyme binding
SUMO binding
methylated histone binding
phosphoprotein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17026298.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026298.hg.1 | ENST00000343048.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026298.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026298.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026301.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026301.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026301.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026302.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026302.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026302.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026299.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026299.hg.1 | ENST00000343048.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026299.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026299.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197642.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197638.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197641.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197639.hg.1 | NM_003655.2 | Refseq |
