Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026296.hg.1 |
ENST00000269397.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026296.hg.1 |
ENST00000343048.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026296.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026296.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026296.hg.1 |
TC17001930.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
TC1700011953.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197636.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000269397.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000448310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000494546.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
ENST00000495122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011953.hg.1 |
NM_003655.2 |
Refseq |