Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013662.hg.1 |
ENST00000243347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013662.hg.1 |
TC02000937.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009662.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
ENST00000460812.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
TC0200009662.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
NM_007115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
ENST00000243347.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167507.hg.1 |
ENST00000460812.1 |
Ensembl |