Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000895.hg.1 |
TC6_dbb_hap3000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3000895.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000952.hg.1 |
TC6_cox_hap2000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2000952.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000933.hg.1 |
TC6_qbl_hap6000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6000933.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003559.hg.1 |
TC06000371.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003559.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003559.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000767.hg.1 |
TC6_mcf_hap5000053.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000767.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000744.hg.1 |
TC6_ssto_hap7000054.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000744.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000908.hg.1 |
TC6_mann_hap4000059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4000908.hg.1 |
NM_000594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151756.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151756.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151756.hg.1 |
TC0600007596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007596.hg.1 |
ENST00000449264.2 |
Ensembl |