Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019661.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019661.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019661.hg.1 |
TC01001279.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010085.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173346.hg.1 |
ENST00000368476.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100173346.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173346.hg.1 |
TC0100010085.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100173346.hg.1 |
NM_000748.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100173346.hg.1 |
ENST00000368476.3 |
Ensembl |