Assay Details
Gene Symbol: CAMSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297707.1 | NP_001284636.1 | 15 - 16 | 4197 | 85 | |
NM_001297708.1 | NP_001284637.1 | 14 - 15 | 4116 | 85 | ||
NM_203459.2 | NP_982284.1 | 14 - 15 | 4164 | 85 | ||
XM_005245041.3 | XP_005245098.1 | 15 - 16 | 4006 | 85 | ||
XM_017000799.1 | XP_016856288.1 | 14 - 15 | 4590 | 85 | ||
GenBank mRNA | AB029001.2 | - | 14 - 15 | 3555 | 85 | |
AL110158.1 | - | 1 - 2 | 77 | 85 | ||
BC011385.1 | - | 4 - 5 | 629 | 85 | ||
BC065508.1 | - | 4 - 5 | 2515 | 85 | ||
BC125229.1 | - | 15 - 16 | 3976 | 40124603 | 85 | |
BC125230.1 | - | 14 - 15 | 3895 | 40124604 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001651.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
