Assay Details
Gene Symbol: CAMSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297708.1 | NP_001284637.1 | 4 - 5 | 917 | 96 | |
NM_203459.2 | NP_982284.1 | 4 - 5 | 917 | 96 | ||
GenBank mRNA | AK095640.1 | - | 4 - 5 | 917 | 96 | |
BC125230.1 | - | 4 - 5 | 696 | 40124604 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
