Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010039.hg.1 |
TC07000643.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010039.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010039.hg.1 |
ENST00000445463.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010039.hg.1 |
ENST00000536888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010039.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008582.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160714.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160714.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160714.hg.1 |
ENST00000223095.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160714.hg.1 |
NM_000602.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160714.hg.1 |
TC0700008582.hg.1 |
Clariom D TC |