Assay Details
Gene Symbol: LEO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286430.1 | NP_001273359.1 | 5 - 6 | 1209 | 156 | |
GenBank mRNA | AK055762.1 | - | 5 - 6 | 1184 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
