Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013370.hg.1 |
ENST00000299601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013370.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013370.hg.1 |
ENST00000538386.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013370.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15013370.hg.1 |
TC15001449.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000558949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000558949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
ENST00000558949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000558949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000558949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
ENST00000558949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009446.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
NM_138792.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
TC1500009446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
ENST00000299601.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
ENST00000315141.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
NM_001286430.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169397.hg.1 |
NM_138792.3 |
Refseq |