Assay Details
Gene Symbol: MIR4290HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038882.1 | 3 - 4 | 1832 | 91 | ||
GenBank mRNA | BC039331.1 | - | 3 - 4 | 1831 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927847 | LOC101927847 | -33.2kb | Anti-sense | Chr. 9 - 89969416 - 89987486 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1508 | LINC01508 | 274.8kb | Sense | Chr. 9 - 90300896 - 90433489 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001315.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809629 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809569 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809596 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149942 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809663 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150049 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149931 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809642 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809623 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149902 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809738 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809755 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150069 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150089 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809637 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150090 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809728 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809699 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150099 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149939 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809670 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809681 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809721 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150061 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809568 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809710 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809643 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150043 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809760 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150086 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809666 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809686 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150006 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150052 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809674 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150074 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149930 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150080 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149897 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809754 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149893 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809622 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809591 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150103 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809586 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150059 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809620 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809684 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150102 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150071 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809651 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150093 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149901 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809638 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809577 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809561 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150084 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149996 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149948 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809608 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149980 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150082 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150027 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809630 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809735 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150035 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149938 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809757 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150039 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150051 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809751 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809709 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809604 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809749 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809726 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150094 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809697 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809680 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150033 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150092 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809647 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150038 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809725 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150001 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809695 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809571 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809633 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809567 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238454 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238450 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809700 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809555 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809723 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150031 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809599 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150058 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809715 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809658 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809741 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809621 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150073 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809610 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809570 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150066 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150097 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16238467 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149976 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809602 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27149978 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809646 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06809566 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27150021 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 129110 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 259873 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 243333 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 305521 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 120498 | BC053850.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 120498 | NM_033260.3 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 96718 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 912970 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 286787 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 407612 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 209059 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3738498 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23489 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3738498 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3738498 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P18689016 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 146910 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P16915081 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02129840 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18689016 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02129822 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P18689016 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02129840 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1561289_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR09016426.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016426.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016426.hg.1 | TC09001315.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900173033.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900173033.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900173033.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900173033.hg.1 | TC0900010723.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900173033.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | ENST00000426593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010723.hg.1 | NR_038882.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1561289_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1561289_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561289_at4 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037675 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037675 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 595706 | - | - |
