Assay Details
Gene Symbol: CAPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145122.1 | NP_001138594.1 | 16 - 17 | 1791 | 72 | |
NM_001321270.1 | NP_001308199.1 | 16 - 17 | 1855 | 72 | ||
XM_011532863.2 | XP_011531165.1 | 17 - 18 | 1952 | 72 | ||
XM_011532864.2 | XP_011531166.1 | 17 - 18 | 1989 | 72 | ||
XM_011532865.1 | XP_011531167.1 | 17 - 18 | 1893 | 72 | ||
XM_011532866.2 | XP_011531168.1 | 15 - 16 | 1660 | 72 | ||
GenBank mRNA | AK092257.1 | - | 16 - 17 | 1756 | 72 | |
AK296910.1 | - | 18 - 19 | 1759 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001715.hg.1
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02027680.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027680.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027680.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027680.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027678.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027678.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027678.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027678.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027681.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027681.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027681.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027681.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02027679.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027679.hg.1 | ENST00000444918.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02027679.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02027679.hg.1 | TC02001715.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188827.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188824.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188826.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | TC0200012168.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200188825.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000398824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | ENST00000403897.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012168.hg.1 | NM_001145122.1 | Refseq |
