Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023406.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023406.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023406.hg.1 |
TC07001558.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023407.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023407.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023407.hg.1 |
TC07001558.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023408.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023408.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023408.hg.1 |
TC07001558.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011624.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182902.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182902.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182902.hg.1 |
TC0700011624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182902.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182902.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182903.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182903.hg.1 |
TC0700011624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182903.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182901.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182901.hg.1 |
NM_001102386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700182901.hg.1 |
TC0700011624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700182901.hg.1 |
ENST00000398291.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700182901.hg.1 |
NM_001102386.2 |
Refseq |