Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000586941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000586941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
ENST00000586941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026321.hg.1 |
ENST00000257572.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026321.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026321.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026321.hg.1 |
TC12002010.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
TC1200012852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200204480.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000550505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000552092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000586941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000586941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
ENST00000586941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NM_003806.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012852.hg.1 |
NR_073189.2 |
Refseq |