Assay Details
Gene Symbol: FUOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098483.2 | NP_001091953.1 | 5 - 6 | 418 | 65 | |
NM_001301827.1 | NP_001288756.1 | 4 - 5 | 349 | 65 | ||
NM_001301828.1 | NP_001288757.1 | 5 - 6 | 625 | 65 | ||
XM_011539672.2 | XP_011537974.1 | 5 - 6 | 457 | 65 | ||
GenBank mRNA | AI333356.1 | - | 1 - 2 | 65 | ||
BM009909.1 | - | 4 - 5 | 281 | 65 | ||
BU849497.1 | - | 5 - 6 | 601 | 65 | ||
BX119427.1 | - | 5 - 6 | 344 | 65 | ||
HY163253.1 | - | 5 - 6 | 413 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001789.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | ENST00000278025.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | ENST00000368552.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | ENST00000447176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | ENST00000465384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023622.hg.1 | TC10001789.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | TC1000012398.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184627.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000278025.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368551.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000368552.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000447176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000465384.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | ENST00000478895.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001098483.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301827.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_001301828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012398.hg.1 | NM_198472.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 174121 | - | - |
