Assay Details
Gene Symbol: CCDC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014008.4 | NP_054727.1 | 11 - 12 | 1496 | 56 | |
XM_005272599.3 | XP_005272656.1 | 11 - 12 | 1425 | 56 | ||
GenBank mRNA | AJ005890.1 | - | 11 - 12 | 1412 | 56 | |
AK298264.1 | - | 11 - 12 | 1491 | 56 | ||
AK316375.1 | - | 11 - 12 | 1442 | 56 | ||
BC000972.2 | - | 11 - 12 | 1443 | 3449051 | 56 | |
BC011675.2 | - | 11 - 12 | 1327 | 4299954 | 56 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000269.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cellular copper ion homeostasis
Golgi to plasma membrane transport
cytoplasmic sequestering of NF-kappaB
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of ubiquitin-dependent protein catabolic process
protein binding
cullin family protein binding
cellular copper ion homeostasis
Golgi to plasma membrane transport
cytoplasmic sequestering of NF-kappaB
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of ubiquitin-dependent protein catabolic process
protein binding
cullin family protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003508.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003508.hg.1 | ENST00000538876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003508.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003508.hg.1 | TC0X000269.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000490300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | ENST00000496651.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007224.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149576.hg.1 | ENST00000376227.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149576.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149576.hg.1 | TC0X00007224.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00149576.hg.1 | NM_014008.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149576.hg.1 | ENST00000376227.3 | Ensembl |
