Assay Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282862.1 | NP_001269791.1 | 9 - 10 | 1142 | 73 | |
NM_145313.3 | NP_660356.2 | 9 - 10 | 1114 | 73 | ||
XM_005271809.3 | XP_005271866.1 | 8 - 9 | 992 | 73 | ||
XM_011539500.2 | XP_011537802.1 | 8 - 9 | 1102 | 73 | ||
GenBank mRNA | AK095136.1 | - | 9 - 10 | 1142 | 73 | |
AK127432.1 | - | 9 - 10 | 3540 | 73 | ||
BC022548.1 | - | 9 - 10 | 1114 | 4814750 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001200.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
