Assay Details
Gene Symbol: ANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001148.4 | NP_001139.3 | 36 - 37 | 4497 | 156 | |
XM_005262945.2 | XP_005263002.1 | 35 - 36 | 4496 | 156 | ||
XM_017008067.1 | XP_016863556.1 | 38 - 39 | 5454 | 156 | ||
XM_017008068.1 | XP_016863557.1 | 39 - 40 | 5695 | 156 | ||
XM_017008069.1 | XP_016863558.1 | 37 - 38 | 5429 | 156 | ||
XM_017008070.1 | XP_016863559.1 | 38 - 39 | 5640 | 156 | ||
XM_017008071.1 | XP_016863560.1 | 39 - 40 | 4623 | 156 | ||
XM_017008072.1 | XP_016863561.1 | 38 - 39 | 4599 | 156 | ||
XM_017008073.1 | XP_016863562.1 | 36 - 37 | 4595 | 156 | ||
XM_017008074.1 | XP_016863563.1 | 38 - 39 | 4569 | 156 | ||
XM_017008075.1 | XP_016863564.1 | 37 - 38 | 4533 | 156 | ||
XM_017008076.1 | XP_016863565.1 | 36 - 37 | 4595 | 156 | ||
XM_017008077.1 | XP_016863566.1 | 35 - 36 | 4410 | 156 | ||
GenBank mRNA | AK299767.1 | - | 37 - 38 | 4521 | 156 | |
AK307601.1 | - | 36 - 37 | 4448 | 156 | ||
Z26634.2 | - | 36 - 37 | 4397 | 156 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
