Assay Details
Gene Symbol: COL5A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015719.3 | NP_056534.2 | 50 - 51 | 3750 | 130 | |
XM_011528042.2 | XP_011526344.1 | 50 - 51 | 3808 | 130 | ||
XM_017026849.1 | XP_016882338.1 | 23 - 24 | 1451 | 130 | ||
GenBank mRNA | AB209175.1 | - | 48 - 49 | 3403 | 130 | |
AF177941.1 | - | 50 - 51 | 3750 | 130 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001156.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
