Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC020770.1 | - | 2 - 3 | 364 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC020770.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
