Assay Details
Gene Symbol: DYNC1LI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286157.1 | NP_001273086.1 | 9 - 10 | 123 | ||
NM_001323955.1 | NP_001310884.1 | 9 - 10 | 123 | |||
NM_006141.2 | NP_006132.1 | 10 - 11 | 123 | |||
NR_136658.1 | 10 - 11 | 123 | ||||
XM_017023008.1 | XP_016878497.1 | 10 - 11 | 123 | |||
GenBank mRNA | AF035812.1 | - | 10 - 11 | 123 | ||
AK095237.1 | - | 3 - 4 | 123 | |||
AK291766.1 | - | 10 - 11 | 123 | |||
AK303031.1 | - | 9 - 10 | 123 | |||
BC025959.1 | - | 10 - 11 | 4329417 | 123 | ||
BU616879.1 | - | 3 - 4 | 475 | 123 | ||
EB387189.1 | - | 7 - 8 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001172.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
