Assay Details
Gene Symbol: ABCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522498.2 | XP_011520800.1 | 12 - 13 | 1735 | 75 | |
GenBank mRNA | HQ917064.1 | - | 1 - 2 | 205 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
