Assay Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024812.2 | NP_079088.1 | 1 - 2 | 365 | 116 | |
GenBank mRNA | AF371319.1 | - | 1 - 2 | 160 | 116 | |
AK022077.1 | - | 1 - 2 | 343 | 116 | ||
BC011517.1 | - | 1 - 2 | 307 | 4183341 | 116 | |
BI548687.1 | - | 1 - 2 | 364 | 114 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000643.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08008747.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08008747.hg.1 | ENST00000309982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08008747.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08008747.hg.1 | TC08000643.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | TC0800008474.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800158151.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
