Assay Details
Gene Symbol: POFUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015352.1 | NP_056167.1 | 3 - 4 | 482 | 78 | |
NM_172236.1 | NP_758436.1 | 3 - 4 | 482 | 78 | ||
GenBank mRNA | AF375884.1 | - | 3 - 4 | 482 | 78 | |
AK291033.1 | - | 3 - 4 | 489 | 78 | ||
AK303068.1 | - | 2 - 3 | 360 | 78 | ||
AL832023.1 | - | 3 - 4 | 414 | 78 | ||
BC000582.2 | - | 3 - 4 | 482 | 3162533 | 78 | |
D80002.2 | - | 3 - 4 | 479 | 78 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000208.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
somitogenesis
fucose metabolic process
regulation of transcription, DNA-templated
protein O-linked glycosylation
Notch signaling pathway
nervous system development
heart development
embryo development
O-glycan processing
protein O-linked fucosylation
fucosyltransferase activity
peptide-O-fucosyltransferase activity
angiogenesis
somitogenesis
fucose metabolic process
regulation of transcription, DNA-templated
protein O-linked glycosylation
Notch signaling pathway
nervous system development
heart development
embryo development
O-glycan processing
protein O-linked fucosylation
fucosyltransferase activity
peptide-O-fucosyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20002562.hg.1 | TC20000208.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002562.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002562.hg.1 | ENST00000375749.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002562.hg.1 | ENST00000486717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002562.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002562.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000408740.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007110.hg.1 | NR_031726.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | TC2000007110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148584.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375730.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000375749.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000408740.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000434904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000465791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | ENST00000486717.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_015352.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NM_172236.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007110.hg.1 | NR_031726.1 | Refseq |
