Assay Details
Gene Symbol: TNFAIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270507.1 | NP_001257436.1 | 1 - 2 | 327 | 69 | |
NM_001270508.1 | NP_001257437.1 | 1 - 2 | 351 | 93 | ||
XM_011536096.1 | XP_011534398.1 | 1 - 2 | 331 | 93 | ||
GenBank mRNA | DB143089.1 | - | 1 - 2 | 297 | 69 | |
HY009939.1 | - | 1 - 2 | 409 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270507.1, NM_001270508.1, XM_011536096.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
