Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012801.hg.1 |
ENST00000260191.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012801.hg.1 |
ENST00000537778.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012801.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11012801.hg.1 |
TC11001024.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009097.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
ENST00000543092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
TC1100009097.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
NM_006028.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
ENST00000260191.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
ENST00000537778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100167443.hg.1 |
ENST00000543092.1 |
Ensembl |