Assay Details
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204118.1 | NP_001191047.1 | 2 - 3 | 201 | 86 | |
NM_207390.3 | NP_997273.3 | 2 - 3 | 201 | 86 | ||
NR_109784.1 | 2 - 3 | 201 | 86 | |||
NR_109785.1 | 2 - 3 | 201 | 86 | |||
XM_017026785.1 | XP_016882274.1 | 2 - 3 | 390 | 86 | ||
XM_017026786.1 | XP_016882275.1 | 2 - 3 | 390 | 86 | ||
XM_017026788.1 | XP_016882277.1 | 2 - 3 | 366 | 86 | ||
XM_017026790.1 | XP_016882279.1 | 2 - 3 | 393 | 86 | ||
XM_017026791.1 | XP_016882280.1 | 2 - 3 | 392 | 86 | ||
XM_017026792.1 | XP_016882281.1 | 2 - 3 | 394 | 86 | ||
XM_017026793.1 | XP_016882282.1 | 2 - 3 | 397 | 86 | ||
XM_017026794.1 | XP_016882283.1 | 2 - 3 | 399 | 86 | ||
GenBank mRNA | BC140848.1 | - | 2 - 3 | 162 | 9021716 | 86 |
BC144664.1 | - | 2 - 3 | 162 | 86 | ||
BC144665.1 | - | 2 - 3 | 162 | 9053196 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000260.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017026785.1, XM_017026786.1, XM_017026788.1, XM_017026790.1, XM_017026791.1, XM_017026792.1, XM_017026793.1, XM_017026794.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
