Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015110.hg.1 |
TC19001101.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015110.hg.1 |
ENST00000245903.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015110.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015107.hg.1 |
TC19001101.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015107.hg.1 |
ENST00000245903.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015107.hg.1 |
ENST00000423145.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015107.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015108.hg.1 |
TC19001101.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015108.hg.1 |
ENST00000245903.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015108.hg.1 |
ENST00000423145.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19015108.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
TC1900009439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175867.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009439.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
TC1900009439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175868.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
TC1900009439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175870.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
TC1900009439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175869.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
NM_001252.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
TC1900009439.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
ENST00000245903.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
ENST00000423145.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
ENST00000597430.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900175866.hg.1 |
NM_001252.4 |
Refseq |