Assay Details
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267713.1 | NP_001254642.1 | 4 - 5 | 726 | 67 | |
NM_002183.3 | NP_002174.1 | 6 - 7 | 960 | 67 | ||
XM_005274431.4 | XP_005274488.1 | 6 - 7 | 997 | 67 | ||
XM_005274432.1 | XP_005274489.1 | 6 - 7 | 787 | 67 | ||
XM_017029491.1 | XP_016884980.1 | 6 - 7 | 995 | 67 | ||
GenBank mRNA | AK290568.1 | - | 6 - 7 | 790 | 67 | |
BC035407.1 | - | 6 - 7 | 960 | 5167281 | 67 | |
FJ550347.1 | - | 3 - 4 | 377 | 67 | ||
M74782.1 | - | 6 - 7 | 757 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | ENST00000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_001267713.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006453.hg.1 | NM_002183.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000331035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000381469.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006452.hg.1 | ENSTR0000432757.5 | Ensembl |
