Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043666.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043666.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043666.hg.1 |
TC02002714.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200214189.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200214189.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200214189.hg.1 |
TC0200015556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200214189.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200214189.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
ENST00000295417.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015556.hg.1 |
NM_003468.3 |
Refseq |