Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001372.hg.1 |
ENST00000330055.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001372.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001372.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04001372.hg.1 |
TC04000052.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
TC0400006616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145883.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006616.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145882.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145882.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145882.hg.1 |
TC0400006616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400145882.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145882.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
NM_000683.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
TC0400006616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
ENST00000330055.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
ENST00000509482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400145884.hg.1 |
NM_000683.3 |
Refseq |