Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004722.3 | NP_001004722.1 | 3 - 4 | 504 | 128 | |
XM_011511992.2 | XP_011510294.2 | 2 - 3 | 448 | 128 | ||
GenBank mRNA | BG480643.1 | - | 3 - 4 | 506 | 129 | |
KU178419.1 | - | 1 - 2 | 228 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
immunological synapse formation
actin filament organization
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
regulation of epidermal growth factor-activated receptor activity
negative regulation of cell population proliferation
vesicle-mediated transport
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
negative regulation of peptidyl-serine phosphorylation
positive regulation of translation in response to endoplasmic reticulum stress
positive regulation of T cell proliferation
T cell activation
positive regulation of GTPase activity
positive regulation of transcription by RNA polymerase II
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
dendritic spine development
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
negative regulation of PERK-mediated unfolded protein response
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
protein binding
cytoskeletal anchor activity
signaling receptor complex adaptor activity
protein-containing complex binding
scaffold protein binding
immunological synapse formation
actin filament organization
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
regulation of epidermal growth factor-activated receptor activity
negative regulation of cell population proliferation
vesicle-mediated transport
cell migration
lamellipodium assembly
positive regulation of actin filament polymerization
negative regulation of peptidyl-serine phosphorylation
positive regulation of translation in response to endoplasmic reticulum stress
positive regulation of T cell proliferation
T cell activation
positive regulation of GTPase activity
positive regulation of transcription by RNA polymerase II
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
dendritic spine development
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
negative regulation of PERK-mediated unfolded protein response
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
protein binding
cytoskeletal anchor activity
signaling receptor complex adaptor activity
protein-containing complex binding
scaffold protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000522586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000522586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | ENST00000522586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000233154.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393348.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000393349.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000425756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000451463.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000522586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000522586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | ENST00000522586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004720.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_001004722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008742.hg.1 | NM_003581.4 | Refseq |
