Assay Details
Gene Symbol: NAA30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011713.2 | NP_001011713.2 | 4 - 5 | 1103 | 141 | |
GenBank mRNA | AK092674.1 | - | 4 - 5 | 1103 | 141 | |
AK296372.1 | - | 3 - 4 | 340 | 141 | ||
AK304257.1 | - | 4 - 5 | 707 | 141 | ||
BC118589.1 | - | 4 - 5 | 1058 | 40085064 | 141 | |
BC122557.1 | - | 5 - 6 | 947 | 40085054 | 141 | |
BX648700.1 | - | 3 - 4 | 384 | 141 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000342.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003655.hg.1 | ENST00000395257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003655.hg.1 | ENST00000555166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003655.hg.1 | ENST00000556492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003655.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003655.hg.1 | TC14000342.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14003656.hg.1 | ENST00000395257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003656.hg.1 | ENST00000556492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003656.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003656.hg.1 | TC14000342.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14003657.hg.1 | ENST00000395257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003657.hg.1 | ENST00000556492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003657.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003657.hg.1 | TC14000342.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14003654.hg.1 | ENST00000395257.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003654.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003654.hg.1 | ENST00000555166.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003654.hg.1 | ENST00000556492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003654.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003654.hg.1 | TC14000342.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | TC1400007285.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150490.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | TC1400007285.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150493.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | TC1400007285.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000554703.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150489.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | TC1400007285.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150492.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | NM_001011713.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | TC1400007285.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | ENST00000298406.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | ENST00000555166.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | ENST00000556492.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150491.hg.1 | NM_001011713.2 | Refseq |
