Assay Details
Gene Symbol: LOC440602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108042.2 | 1 - 2 | 123 | 101 | ||
GenBank mRNA | BC042100.1 | - | 1 - 2 | 123 | 101 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BQ049505.1 | - | 121 | Chr. - - [-] on Build | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chymosin pseudogene | CYMP | Anti-sense | Chr. 1 - 110480766 - 110491269 on NCBI Build GRCh38 | ||
Downstream | potassium voltage-gated channel subfamily A member 10 | KCNA10 |
Kcn1, Kv1.8 |
27.0kb | Sense | Chr. 1 - 110517217 - 110519175 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454509 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454458 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487143 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487161 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454474 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454508 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454543 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454523 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454490 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487152 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454476 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454497 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454518 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454510 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487170 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487133 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454467 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487115 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454535 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487121 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487172 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487165 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454473 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454538 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454487 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487136 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454505 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454527 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454496 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487183 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487150 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487117 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454548 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454541 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454482 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487173 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487137 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454501 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454516 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20487186 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00454534 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3787645 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29395 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3787645 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35267606 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1561335_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100015241.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015241.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015241.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015241.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213622.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213622.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213622.hg.1 | TC0100015241.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100213622.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213622.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015241.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015241.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015241.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015241.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213621.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213621.hg.1 | NR_108042.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213621.hg.1 | TC0100015241.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100213621.hg.1 | ENST00000457402.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213621.hg.1 | NR_108042.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 1561335_at | - | - |
Human Genome U133 Plus 2.0 Array | 1561335_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1561335_at11 | - | - |
