Assay Details
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK123528.1 | - | 1 | 356 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | rabphilin 3A | RPH3A | -16.7kb | Sense | Chr. 12 - 112791538 - 112898881 on NCBI Build GRCh38 | |
Downstream | 2'-5'-oligoadenylate synthetase 3 | OAS3 |
p100, p100OAS |
20.5kb | Sense | Chr. 12 - 112938433 - 112973251 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
