Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000488874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000488874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
ENST00000488874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007416.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006125.hg.1 |
TC22000322.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22006125.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000473717.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000488874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000488874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
ENST00000488874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NM_138435.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007416.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152359.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152359.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152359.hg.1 |
TC2200007416.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152359.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152359.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152360.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152360.hg.1 |
NR_039988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152360.hg.1 |
TC2200007416.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152360.hg.1 |
ENST00000333407.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152360.hg.1 |
NR_039988.1 |
Refseq |
Human Genome U95 Set |
52480_at |
- |
- |
Human Genome U95 Set |
52480_PM_at |
- |
- |