Assay Details
Gene Symbol: GPM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127103.1 | - | 1 | 3146 | 56 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
OFD1, centriole and centriolar satellite protein |
OFD1 |
71-7A, CXorf5, JBTS10, RP23, SGBS2 |
-55.9kb | Anti-sense | Chr. X - 13734713 - 13769361 on NCBI Build GRCh38 |
Downstream | gem nuclear organelle associated protein 8 | GEMIN8 | FAM51A1 | 171.4kb | Sense | Chr. X - 14002494 - 14030087 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
