Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000459421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000459421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000490661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000490661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000490661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000497592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
ENST00000497592.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NR_031747.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011892.hg.1 |
NR_031747.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008595.hg.1 |
ENST00000624170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008595.hg.1 |
ENST00000624170.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008595.hg.1 |
ENST00000624170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008595.hg.1 |
ENST00000624170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008595.hg.1 |
ENST00000624170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008595.hg.1 |
ENST00000624170.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000369287.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000428953.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000459421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000459421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000490661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000490661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000490661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000497592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
ENST00000497592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NM_018017.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NR_031747.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011892.hg.1 |
NR_031747.1 |
Refseq |