Assay Details
Gene Symbol: GLI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB209654.1 | - | 2 | 4251 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZFP41 zinc finger protein | ZFP41 |
ZNF753, zfp-41 |
-11.0kb | Sense | Chr. 8 - 143246821 - 143262705 on NCBI Build GRCh38 |
Downstream | MYC-induced long noncoding RNA | MINCR | LINC01604 | 6.3kb | Anti-sense | Chr. 8 - 143280155 - 143281700 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000340042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000344692.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517468.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000517530.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000519876.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000520021.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000521682.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522033.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000522479.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | ENST00000523812.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012374.hg.1 | NM_138465.3 | Refseq |
