Assay Details
Gene Symbol: LOC283745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK097986.1 | - | 1 | 346 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carbohydrate sulfotransferase 14 | CHST14 |
ATCS, D4ST1, EDSMC1, HNK1ST |
-54.3kb | Anti-sense | Chr. 15 - 40470961 - 40473158 on NCBI Build GRCh38 |
Downstream | chromosome 15 open reading frame 57 | C15orf57 | CCDC32 | 2.8kb | Sense | Chr. 15 - 40531341 - 40565057 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
