Assay Details
Gene Symbol: GOLGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX640663.1 | - | 1 | 1948 | 93 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AW301254.1 | - | 1 | Chr. 3 - 37360943 - 37361429 [-] on Build GRCh38 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LRR binding FLII interacting protein 2 | LRRFIP2 | HUFI-2 | -182.6kb | Anti-sense | Chr. 3 - 37052626 - 37176817 on NCBI Build GRCh38 |
Downstream | chromosome 3 open reading frame 35 | C3orf35 | APRG1 | 23.9kb | Sense | Chr. 3 - 37379111 - 37435497 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
