Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011020.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011020.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011020.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011020.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011020.hg.1 |
TC04000800.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011018.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011018.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011018.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011018.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011018.hg.1 |
TC04000800.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011017.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011017.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011017.hg.1 |
TC04000800.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
TC0400009112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162655.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
TC0400009112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162653.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
TC0400009112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162652.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000455598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000507108.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
ENST00000509450.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009112.hg.1 |
NR_026992.1 |
Refseq |