Assay Details
Gene Symbol: TISP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027314.1 | 1 - 2 | 139 | 76 | ||
GenBank mRNA | BC037249.1 | - | 1 - 2 | 139 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC646743 | LOC646743 | -20.9kb | Anti-sense | Chr. 2 - 130536570 - 130549915 on NCBI Build GRCh38 | |
Downstream |
cripto, FRL-1, cryptic family 1 |
CFC1 |
CFC1B, CRYPTIC, DTGA2, HTX2 |
8.0kb | Anti-sense | Chr. 2 - 130592165 - 130599575 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000815.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02012547.hg.1 | ENST00000409793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012547.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012547.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012547.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012547.hg.1 | TC02000815.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200009306.hg.1 | ENST00000422631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009306.hg.1 | ENST00000422631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012557.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012557.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012557.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012557.hg.1 | TC02000815.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200014275.hg.1 | ENST00000457169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014275.hg.1 | ENST00000457169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012559.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012559.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012559.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012559.hg.1 | TC02000815.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012548.hg.1 | ENST00000409793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012548.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02012548.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012548.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012548.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012548.hg.1 | TC02000815.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02012558.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012558.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012558.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02012558.hg.1 | TC02000815.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | TC0200014273.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200204314.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009306.hg.1 | ENST00000422631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009306.hg.1 | ENST00000422631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014275.hg.1 | ENST00000457169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014275.hg.1 | ENST00000457169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409793.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000409982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000414595.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000419965.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000440359.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009307.hg.1 | ENST00000623376.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | ENST00000409259.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_027314.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_033930.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014273.hg.1 | NR_033930.1 | Refseq |
